EMAIL THIS PAGE TO A FRIEND
Applied immunohistochemistry & molecular morphology : AIMM
Combination of napsin A and TTF-1 immunohistochemistry helps in differentiating primary lung adenocarcinoma from metastatic carcinoma in the lung.
PMID 21464700
Abstract
Differentiation of primary from metastatic adenocarcinoma in the lung can be challenging, and it demands sensitive and specific biomarkers, especially when the tissue for diagnosis is limited. Thyroid transcription factor-1 (TTF-1) has been considered a reliable marker for adenocarcinoma of lung origin. However, several recent studies have shown that TTF-1 immunostaining is also positive in adenocarcinomas arising in different organs including colon, endometrium, endocervix, and ovary. In addition, approximately 20% of lung primary adenocarcinomas are negative for TTF-1 immunostaining, and napsin A immunostaining has slightly higher sensitivity in detecting lung primary adenocarcinoma. We performed TTF-1 and napsin A immunostaining on 120 cases of primary lung adenocarcinomas and 37 cases of metastatic carcinomas in the lung. The results showed that 95 (79.2%) of 120 lung primary adenocarcinomas showed napsin A+/TTF-1+ double-positive immunostaining pattern. TTF-1⁻/napsin A+, TTF-1+/napsin A⁻, and TTF-1⁻/napsin A⁻ were seen in 8.3%, 3.3%, and 9.2% lung primary adenocarcinomas, respectively. Eight (21.6%) of the 37 metastatic carcinomas were positive for TTF-1 and they include clear-cell renal cell carcinomas completely negative for napsin A although napsin A was detected in 12 (80.0%) of 15 primary papillary and 3 (33.3%) of 9 primary clear-cell renal cell carcinomas. All renal epithelial neoplasms were TTF-1 negative. These findings indicate that double napsin A and TTF-1-positive immunostaining is highly specific for lung primary adenocarcinoma and the combination of these 2 biomarkers is warranted to help segregating primary lung adenocarcinoma from metastatic carcinoma in the lung.