EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
High-performance air-stable organic field-effect transistors: isoindigo-based conjugated polymers.
PMID 21466199
Abstract
Two conjugated polymers, IIDDT and IIDT, based on an isoindigo core were developed for organic field-effect transisitors. Investigation of their field-effect performance indicated that IIDDT exhibited air-stable mobility up to 0.79 cm(2) V(-1) s(-1), which is quite high among polymer FET materials. The facile preparation and high mobility of such polymers make isoindigo-based polymers very promising for application as solution-processable organic semiconductors for optoelectronic devices.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
802123
6,6′-Dibromoisoindigo
C16H8Br2N2O2