EMAIL THIS PAGE TO A FRIEND
Current protocols in molecular biology
Construction of normalized RNA-seq libraries for next-generation sequencing using the crab duplex-specific nuclease.
PMID 21472699
Abstract
RNA-seq is a method for studying the transcriptome of cells or tissues by massively parallel sequencing of tens of millions of short DNA fragments. However, the broad dynamic range of gene expression levels, which span more than five orders of magnitude, necessitates considerable over-sequencing to characterize low-abundance RNAs at sufficient depth. Here, we describe a method that enables efficient sequencing of low-abundance RNAs by normalizing or reducing the range spanned by the most abundant RNA species to the least abundant RNA species. This normalization is achieved using an approach that was developed for generating expressed sequence tag (EST) libraries that uses the crab duplex-specific nuclease and exploits the kinetics of DNA annealing. That is, double-stranded cDNA is denatured, then allowed to partially re-anneal, and the most abundant species, which re-anneal most rapidly, are digested with crab duplex-specific nuclease. This procedure substantially decreases the proportion of sequence reads from highly expressed RNAs, facilitating assessment of the full spectrum of the sequence and structure of transcriptomes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
DATP-RO
dATP, PCR Grade, sodium salt