EMAIL THIS PAGE TO A FRIEND
Nihon yakurigaku zasshi. Folia pharmacologica Japonica
[Novel anti-cancer effects of indomethacin: exploring the cyclooxygenase-inhibition-independent effects].
PMID 21478637