EMAIL THIS PAGE TO A FRIEND
Journal of visualized experiments : JoVE
Using whole mount in situ hybridization to link molecular and organismal biology.
PMID 21490578
Abstract
Whole mount in situ hybridization (WISH) is a common technique in molecular biology laboratories used to study gene expression through the localization of specific mRNA transcripts within whole mount specimen. This technique (adapted from Albertson and Yelick, 2005) was used in an upper level undergraduate Comparative Vertebrate Biology laboratory classroom at Syracuse University. The first two thirds of the Comparative Vertebrate Biology lab course gave students the opportunity to study the embryology and gross anatomy of several organisms representing various chordate taxa primarily via traditional dissections and the use of models. The final portion of the course involved an innovative approach to teaching anatomy through observation of vertebrate development employing molecular techniques in which WISH was performed on zebrafish embryos. A heterozygous fibroblast growth factor 8 a (fgf8a) mutant line, ace, was used. Due to Mendelian inheritance, ace intercrosses produced wild type, heterozygous, and homozygous ace/fgf8a mutants in a 1:2:1 ratio. RNA probes with known expression patterns in the midline and in developing anatomical structures such as the heart, somites, tailbud, myotome, and brain were used. WISH was performed using zebrafish at the 13 somite and prim-6 stages, with students performing the staining reaction in class. The study of zebrafish embryos at different stages of development gave students the ability to observe how these anatomical structures changed over ontogeny. In addition, some ace/fgf8a mutants displayed improper heart looping, and defects in somite and brain development. The students in this lab observed the normal development of various organ systems using both external anatomy as well as gene expression patterns. They also identified and described embryos displaying improper anatomical development and gene expression (i.e., putative mutants). For instructors at institutions that do not already own the necessary equipment or where funds for lab and curricular innovation are limited, the financial cost of the reagents and apparatus may be a factor to consider, as will the time and effort required on the part of the instructor regardless of the setting. Nevertheless, we contend that the use of WISH in this type of classroom laboratory setting can provide an important link between developmental genetics and anatomy. As technology advances and the ability to study organismal development at the molecular level becomes easier, cheaper, and increasingly popular, many evolutionary biologists, ecologists, and physiologists are turning to research strategies in the field of molecular biology. Using WISH in a Comparative Vertebrate Biology laboratory classroom is one example of how molecules and anatomy can converge within a single course. This gives upper level college students the opportunity to practice modern biological research techniques, leading to a more diversified education and the promotion of future interdisciplinary scientific research.
Related Materials