EMAIL THIS PAGE TO A FRIEND
Hellenic journal of nuclear medicine
Alternative production methods to face global molybdenum-99 supply shortage.
PMID 21512666
Abstract
The sleeping giant of molybdenum-99 ((99)Mo) production is grinding to a halt and the world is wondering how this happened. Fewer than 10 reactors in the world are capable of producing radio nuclides for medicine; approximately 50% of the world's supply of raw material comes from National Research Universal (NRU) reactor in Canada. Many of these reactors, like the NRU, are old and aging. No one of these reactors, and probably not even all of them in combination, can replace the production of NRU. As the healthcare industry faces an aging population and the demand for diagnostic services using (99m)Tc continues to rise, the need for a consistent, reliable supply of (99)Mo has become increasingly important, so alternative methods to produce (99)Mo or even directly (99m)Tc had to be considered to avoid a supply shortage in the coming years. This need guides to the production of (99)Mo by replacing the Highly Enriched Uranium (HEU) target in a nuclear reactor with Low Enriched Uranium (LEU) and furthermore to the use of accelerators for manufacturing (99)Mo or for directly producing (99m)Tc.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF42672427
Molybdenum, foil, not light tested, 25x25mm, thickness 0.003mm, 99.9%
Mo
GF11484750
Molybdenum, foil, not light tested, 25x25mm, thickness 0.01mm, 99.9%
Mo
GF81739457
Molybdenum, foil, not light tested, 50x50mm, thickness 0.003mm, 99.9%
Mo
GF30193781
Molybdenum, foil, not light tested, 50x50mm, thickness 0.0125mm, 99.9%
Mo
GF56008986
Molybdenum, insulated wire, 5m, conductor diameter 0.125mm, insulation thickness 0.012mm, polyimide insulation, 99.95%
Mo
GF28017637
Molybdenum, microfoil, disks, 10mm, thinness 0.1μm, specific density 92.4μg/cm2, permanent mylar 3.5μm support, 99.9%
Mo
GF04272049
Molybdenum, pellets, 200g, max. size 10mm, 99.9%
Mo
GF54490574
Molybdenum, rod, 1000mm, diameter 2.0mm, centerless ground, 99.9%
Mo
GF60430766
Molybdenum, rod, 100mm, diameter 1.25mm, centerless ground, 99.9%
Mo
GF11351359
Molybdenum, wire, straight, 500mm, diameter 1.0mm, 99.95%
Mo
GF66431606
Molybdenum, foil, not light tested, 25x25mm, thickness 0.006mm, 99.9%
Mo
GF23412383
Molybdenum, foil, not light tested, 25x25mm, thickness 0.008mm, 99.9%
Mo
GF90888788
Molybdenum, foil, not light tested, 25x25mm, thickness 0.0125mm, 99.9%
Mo
GF98811355
Molybdenum, foil, not light tested, 25x25mm, thickness 0.01mm, annealed, 99.9%
Mo
GF26098027
Molybdenum, foil, not light tested, 50x50mm, thickness 0.001mm, 99.9%
Mo
GF63646218
Molybdenum, foil, not light tested, 50x50mm, thickness 0.0025mm, 99.9%
Mo
GF20045786
Molybdenum, foil, not light tested, 50x50mm, thickness 0.002mm, 99.9%
Mo
GF47495975
Molybdenum, foil, not light tested, 50x50mm, thickness 0.004mm, 99.9%
Mo
GF48792628
Molybdenum, foil, not light tested, 50x50mm, thickness 0.005mm, 99.9%
Mo
GF74350267
Molybdenum, foil, not light tested, 50x50mm, thickness 0.006mm, 99.9%
Mo
GF37145358
Molybdenum, foil, not light tested, 50x50mm, thickness 0.007mm, 99.9%
Mo
GF95631419
Molybdenum, foil, not light tested, 50x50mm, thickness 0.008mm, 99.9%
Mo
GF41758768
Molybdenum, foil, not light tested, 50x50mm, thickness 0.009mm, 99.9%
Mo
GF99294968
Molybdenum, foil, not light tested, 50x50mm, thickness 0.01mm, 99.9%
Mo
GF42252016
Molybdenum, foil, not light tested, 50x50mm, thickness 0.01mm, annealed, 99.9%
Mo
GF01123489
Molybdenum, insulated wire, 10m, conductor diameter 0.125mm, insulation thickness 0.012mm, polyimide insulation, 99.95%
Mo
GF40435972
Molybdenum, insulated wire, 20m, conductor diameter 0.125mm, insulation thickness 0.012mm, polyimide insulation, 99.95%
Mo
GF54497111
Molybdenum, insulated wire, 50m, conductor diameter 0.125mm, insulation thickness 0.012mm, polyimide insulation, 99.95%
Mo
GF33272333
Molybdenum, mesh, 100x100mm, nominal aperture 0.44mm, wire diameter 0.07mm, 50 wires/inch, open area 67%, plain weave mesh
Mo
GF01979477
Molybdenum, mesh, 100x100mm, nominal aperture 0.8mm, wire diameter 0.18mm, 26 wires/inch, open area 67%, plain weave mesh
Mo