EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis and SAR study of tricyclic sulfones as γ-secretase inhibitors: C-6 and C-8 positions.
PMID 21515050
Abstract
SAR exploration at C-6 and C-8 positions of the tricyclic sulfone series was carried out. Several functional groups were found to be well tolerated at C-6 and C-8 positions. Selective combination of C-6 and C-8 modification resulted in new tricyclic sulfone analogs with efficacy in in vivo mouse Aβ(40) lowering model.