EMAIL THIS PAGE TO A FRIEND
Apoptosis : an international journal on programmed cell death
Apoptosis induced by overall metabolic stress converges on the Bcl-2 family proteins Noxa and Mcl-1.
PMID 21516346
Abstract
Apoptosis provoked by glucose shortage in dividing T cells is mediated via the BH3-only protein Noxa and inhibition of its binding partner Mcl-1. It is unknown how signals from cellular metabolism can affect the balance between Mcl-1 and Noxa and to what extent other Bcl-2 members are involved in this apoptosis cascade. Here, we defined the mechanism underlying apoptosis in relation to various types of metabolic stress. First, we established that the Noxa/Mcl-1 balance is regulated by glucose deprivation as well as by general metabolic stress, via changes in proteasome-mediated degradation of Mcl-1. Second, in contrast with cytokine-deprivation, no transcriptional modulation of Mcl-1, Puma, Bim or Noxa was observed during glucose deprivation. Third, no changes in PKB or GSK3 activity occurred and no clear role for AMPK was detected. Fourth, apoptosis triggered by nutrient deprivation was executed without signs of overt autophagy and independent of ROS production or p38 MAP kinase activity. Lastly, apoptosis under nutrient limitation could also be delayed by knock-down of Bim or overexpression of Bcl-2. In conclusion, Noxa functions in a specific apoptotic pathway that integrates overall nutrient stress, independent from attenuated PI3K/PKB signaling and without clear involvement of autophagy.