EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Two-step synthesis of novel, bioactive derivatives of the ubiquitous cofactor nicotinamide adenine dinucleotide (NAD).
PMID 21528845
Abstract
We report the design and concise synthesis, in two steps from commercially available material, of novel, bioactive derivatives of the enzyme cofactor nicotinamide adenine dinucleotide (NAD). The new synthetic dinucleotides act as sirtuin (SIRT) inhibitors and show isoform selectivity for SIRT2 over SIRT1. An NMR-based conformational analysis suggests that the conformational preferences of individual analogues may contribute to their isoform selectivity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B6272
8-Bromoadenosine, powder
C10H12BrN5O4