EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Highly potent activity of (1R,2R,6S)-3-methyl-6-(prop-1-en-2-yl)cyclohex-3-ene-1,2-diol in animal models of Parkinson's disease.
PMID 21534547
Abstract
(1R,2R,6S)-3-Methyl-6-(prop-1-en-2-yl)cyclohex-3-ene-1,2-diol 1 possesses potent antiparkinsonian activity in both MPTP and haloperidol animal models. The use of compound 1 resulted in nearly full recovery of the locomotor and exploratory activities and was as effective as the comparator agent (levodopa). All eight stereoisomers of compound 1 have been synthesized and the influence of the absolute configuration on the antiparkinsonian activity of compound 1 was shown.