EMAIL THIS PAGE TO A FRIEND
Brain injury
Adverse cognitive effects of phenytoin in severe brain injury: a case report.
PMID 21534742
Abstract
The use of prophylactic anticonvulsants following brain injury is controversial. When used for this reason or for treatment of early seizures, anticonvulsants, particularly phenytoin, can cause severe cognitive side-effects. This study presents a case of a woman with a severe brain injury with severe cognitive impairment who improved dramatically following withdrawal of phenytoin. The literature regarding such cognitive side-effects is contradictory with no consistent indication of choice of anticonvulsants to use in this situation. As a result of the dramatic improvement in this case, one should now routinely withdraw or change phenytoin treatment in all brain injury patients with significant cognitive impairment.
Related Materials