EMAIL THIS PAGE TO A FRIEND
Acta psychiatrica Scandinavica
The search of new biomarkers to identify Alzheimer's disease: an editorial comment to T. Vanmierlo et al. 'The plant sterol brassicasterol and additional CFS biomarker in Alzheimer's Disease' (1).
PMID 21564039
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B4936
Brassicasterol, from semisynthetic
C28H46O