EMAIL THIS PAGE TO A FRIEND
Acta crystallographica. Section E, Structure reports online
2-Chloro-pyridine-3-carboxamide.
PMID 21581971
Abstract
In the crystal structure of the title compound, C(6)H(5)ClN(2)O, the dihedral angle between the pyridine ring and the carboxamine group is 63.88 (8)°. Inter-molecular N-H⋯N and N-H⋯O hydrogen bonds link the mol-ecules into a two-dimensional network.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
458821
2-Chloropyridine-3-carbonyl chloride, 98%
C6H3Cl2NO