EMAIL THIS PAGE TO A FRIEND
British journal of cancer
Promoter methylation of Wnt5a is associated with microsatellite instability and BRAF V600E mutation in two large populations of colorectal cancer patients.
PMID 21587258
Abstract
In colorectal cancer (CRC), tumour microsatellite instability (MSI) status and CpG island methylator phenotype (CIMP) status are indicators of patient outcome, but the molecular events that give rise to these outcomes remain largely unknown. Wnt5a is a critical regulator of non-canonical Wnt activity and promoter hypermethylation of this gene has emerging prognostic roles in CRC; however the frequency and prognostic significance of this epigenetic event have not been explored in the context of colorectal tumour subtype. Consequently, we investigated the frequency and prognostic significance of Wnt5a methylation in a large cohort of MSI-stratified CRCs. Methylation was quantified in a large cohort of 1232 colorectal carcinomas from two clinically distinct populations from Canada. Associations were examined between methylation status and clinicopathlogical features, including tumour MSI status, BRAF V600E mutation, and patient survival. In Ontario, Wnt5a methylation was strongly associated with MSI tumours after adjustment for age, sex, and tumour location (odds ratio (OR)=4.2, 95% confidence interval (CI)=2.4-7.4, P<10(-6)) and with BRAF V600E mutation, a marker of CIMP (OR=12.3, 95% CI=6.9-21.7, P<10(-17)), but was not associated with patient survival. Concordant results were obtained in Newfoundland. Methylation of Wnt5a is associated with distinct tumour subtypes, strengthening the evidence of an epigenetic-mediated Wnt bias in CRC.