EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Inhibition of periarterial nerve stimulation-induced vasodilation of the mesenteric arterial bed by CGRP (8-37) and CGRP receptor desensitization.
PMID 2159300
Abstract
We provide the first functional evidence that calcitonin gene-related peptide (8-37) induces a direct vasoconstriction and reversibly antagonizes vasodilation of the mesenteric arterial bed induced by calcitonin gene-related peptide (CGRP) suggesting that CGRP (8-37) is a competitive antagonist of vascular CGRP receptors. Vasodilation induced by periarterial nerve stimulation was inhibited both by CGRP (8-37) and by desensitization of CGRP receptors. These results further support the evidence that the periarterial nerve stimulation-induced nonadrenergic noncholinergic vasodilation of the mesenteric vasculature is mediated by endogenous CGRP and its receptors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C2806
Calcitonin Gene Related Peptide Fragment 8-37 human, ≥97% (HPLC), powder
C139H230N44O38