EMAIL THIS PAGE TO A FRIEND
Enzyme research
Synthesis of isopropyl ferulate using silica-immobilized lipase in an organic medium.
PMID 21603272
Abstract
Immobilization of lipases has proved to be a useful technique for improving an enzyme's activity in organic solvents. In the present study, the performance of a silica-immobilized lipase was evaluated for the synthesis of isopropyl ferulate in DMSO. The biocatalyst was cross-linked onto the matrix with 1% glutaraldehyde. The effects of various parameters, molar ratio of ferulic acid to isopropyl alcohol (25 mM : 100 mM), concentration of biocatalyst (2.5-20 mg/mL), molecular sieves (25-250 mg/mL), and various salt ions, were studied consecutively as a function of percent esterification. Immobilized lipase at 25 mg/mL showed maximum esterification (~84%) of ferulic acid and isopropanol at a molar ratio of 25 mM : 100 mM, respectively, in DMSO at 45°C in 3 h under shaking (150 rpm). To overcome the inhibitory effect of water (a byproduct) if any, in the reaction mixture, molecular sieves (3 Å × 1.5 mm; 100 mg/mL) were added to the reaction mixture to promote the forward reaction. Salt ions like Ca(2+), Cd(2+), and Fe(2+) enhanced the activity of immobilized biocatalyst while a few ions like Co(2+), Zn(2+), Mg(2+), Mn(2+), Al(3+), and Na(+) had mild inhibitory effect. Approximately, one third of total decrease in the esterification efficacy was observed after the 5th repetitive cycle of esterification.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
90034
4-Hydroxy-3-methoxycinnamic acid, mixture of isomers, analytical reference material
C10H10O4