EMAIL THIS PAGE TO A FRIEND
Applied Catalysis A: General
Determination of adsorption and kinetic parameters for methyl acetate esterification and hydrolysis reaction catalyzed by Amberlyst 15
PMID 216380