EMAIL THIS PAGE TO A FRIEND
Chemical Society reviews
Palladium-catalysed hydroxylation and alkoxylation.
PMID 21643619
Abstract
The formation of oxygen-carbon bonds is one of the fundamental transformations in organic synthesis. In this regard the application of palladium-based catalysts has been extensively studied during recent years. Nowadays it is an established methodology and the success has been proven in manifold synthetic procedures. This tutorial review summarizes the advances on palladium-catalysed C-O bond formation, means hydroxylation and alkoxylation reactions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
373192
Palladium, evaporation slug, diam. × L 0.9 cm × 1.2 cm, 99.95% trace metals basis
Pd
373206
Palladium, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.95% trace metals basis
Pd
348678
Palladium, foil, thickness 1.0 mm, 99.9% trace metals basis
Pd
287474
Palladium, foil, thickness 0.5 mm, 99.9% trace metals basis
Pd
348643
Palladium, foil, thickness 0.25 mm, 99.98% trace metals basis
Pd
411450
Palladium, foil, thickness 0.05 mm, 99.9% trace metals basis
Pd
267120
Palladium, foil, thickness 0.025 mm, 99.9% trace metals basis
Pd
203998
Palladium, granular, 99.99% trace metals basis
Pd
203939
Palladium, powder, 99.999% trace metals basis
Pd
464651
Palladium, powder, <75 μm, 99.9% trace metals basis
Pd
326666
Palladium, powder, <1 μm, ≥99.9% trace metals basis
Pd
346985
Palladium, rod, diam. 6.25 mm, ≥99.9% trace metals basis
Pd
267082
Palladium, sponge, 99.9% trace metals basis
Pd
267112
Palladium, wire, diam. 1.0 mm, 99.9% trace metals basis
Pd
348694
Palladium, wire, diam. 0.5 mm, 99.9% trace metals basis
Pd
643181
Palladium, extent of labeling: 5 wt. % loading (dry basis), matrix activated carbon, wet support, Degussa type E105CA/W
Pd
686468
Palladium, nanopowder, <25 nm particle size (TEM), ≥99.5%
Pd
GF46569831
Palladium, foil, 6mm disks, thickness 0.025mm, as rolled, 99.99+%
Pd
GF29399158
Palladium, foil, 6mm disks, thickness 0.25mm, as rolled, 99.95%
Pd
GF21228825
Palladium, foil, 8mm disks, thickness 0.025mm, as rolled, 99.99+%
Pd
GF79862833
Palladium, wire reel, 0.05m, diameter 1.0mm, as drawn, 99.99+%
Pd
GF22660264
Palladium, wire reel, 0.1m, diameter 0.5mm, as drawn, 99.95%
Pd
GF00436733
Palladium, wire reel, 0.5m, diameter 1.0mm, as drawn, 99.99+%
Pd
GF85707513
Palladium, wire reel, 0.5m, diameter 1.0mm, hard, 99.95%
Pd
GF76572940
Palladium, wire reel, 1m, diameter 0.025mm, as drawn, 99.9%
Pd
GF96794409
Palladium, wire reel, 25m, diameter 0.125mm, hard, 99.95%
Pd
GF58966037
Palladium, wire reel, 2m, diameter 0.5mm, as drawn, 99.95%
Pd
GF24288790
Palladium, wire reel, 5m, diameter 0.25mm, hard, 99.95%
Pd
GF02178761
Palladium, wire reel, 2m, diameter 0.025mm, as drawn, 99.9%
Pd
GF93105547
Palladium, wire reel, 2m, diameter 0.02mm, as drawn, 99.95%
Pd