EMAIL THIS PAGE TO A FRIEND
Journal of cellular biochemistry
Molybdenum enzymes and molybdenum cofactor in mycobacteria.
PMID 21678480
Abstract
When intracelluar pathogens enter the host macrophages where in addition to oxidative and antibiotic mechanisms of antimicrobial activity, nutrients are deprived. Human pathogen Mycobacterium tuberculosis is one of macrophage parasitisms, which can replicate and persist for decades in dormancy state in virulent environments. It is very successful in escaping the killing mechanisms of macrophage. Molybdenum (Mo) enzymes involve in the global carbon, sulfur, and nitrogen cycles by catalyzing important redox reactions. There are several Mo enzymes in mycobacteria and they exert several important physiological functions, such as dormancy regulation, the metabolism of energy sources, and nitrogen source. Pterin-based Mo cofactor (Moco) is the common cofactor of the Mo enzymes in mycobacteria but the cofactor biosynthesis is nearly an untapped area. The present article discusses the physiological function of Mo enzymes and the structural feature of the genes coding for Moco biosynthesis enzymes in mycobacteria.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF42672427
Molybdenum, foil, not light tested, 25x25mm, thickness 0.003mm, 99.9%
Mo
GF11484750
Molybdenum, foil, not light tested, 25x25mm, thickness 0.01mm, 99.9%
Mo
GF81739457
Molybdenum, foil, not light tested, 50x50mm, thickness 0.003mm, 99.9%
Mo
GF30193781
Molybdenum, foil, not light tested, 50x50mm, thickness 0.0125mm, 99.9%
Mo
GF56008986
Molybdenum, insulated wire, 5m, conductor diameter 0.125mm, insulation thickness 0.012mm, polyimide insulation, 99.95%
Mo
GF28017637
Molybdenum, microfoil, disks, 10mm, thinness 0.1μm, specific density 92.4μg/cm2, permanent mylar 3.5μm support, 99.9%
Mo
GF04272049
Molybdenum, pellets, 200g, max. size 10mm, 99.9%
Mo
GF54490574
Molybdenum, rod, 1000mm, diameter 2.0mm, centerless ground, 99.9%
Mo
GF60430766
Molybdenum, rod, 100mm, diameter 1.25mm, centerless ground, 99.9%
Mo
GF11351359
Molybdenum, wire, straight, 500mm, diameter 1.0mm, 99.95%
Mo
GF66431606
Molybdenum, foil, not light tested, 25x25mm, thickness 0.006mm, 99.9%
Mo
GF23412383
Molybdenum, foil, not light tested, 25x25mm, thickness 0.008mm, 99.9%
Mo
GF90888788
Molybdenum, foil, not light tested, 25x25mm, thickness 0.0125mm, 99.9%
Mo
GF98811355
Molybdenum, foil, not light tested, 25x25mm, thickness 0.01mm, annealed, 99.9%
Mo
GF26098027
Molybdenum, foil, not light tested, 50x50mm, thickness 0.001mm, 99.9%
Mo
GF63646218
Molybdenum, foil, not light tested, 50x50mm, thickness 0.0025mm, 99.9%
Mo
GF20045786
Molybdenum, foil, not light tested, 50x50mm, thickness 0.002mm, 99.9%
Mo
GF47495975
Molybdenum, foil, not light tested, 50x50mm, thickness 0.004mm, 99.9%
Mo
GF48792628
Molybdenum, foil, not light tested, 50x50mm, thickness 0.005mm, 99.9%
Mo
GF74350267
Molybdenum, foil, not light tested, 50x50mm, thickness 0.006mm, 99.9%
Mo
GF37145358
Molybdenum, foil, not light tested, 50x50mm, thickness 0.007mm, 99.9%
Mo
GF95631419
Molybdenum, foil, not light tested, 50x50mm, thickness 0.008mm, 99.9%
Mo
GF41758768
Molybdenum, foil, not light tested, 50x50mm, thickness 0.009mm, 99.9%
Mo
GF99294968
Molybdenum, foil, not light tested, 50x50mm, thickness 0.01mm, 99.9%
Mo
GF42252016
Molybdenum, foil, not light tested, 50x50mm, thickness 0.01mm, annealed, 99.9%
Mo
GF01123489
Molybdenum, insulated wire, 10m, conductor diameter 0.125mm, insulation thickness 0.012mm, polyimide insulation, 99.95%
Mo
GF40435972
Molybdenum, insulated wire, 20m, conductor diameter 0.125mm, insulation thickness 0.012mm, polyimide insulation, 99.95%
Mo
GF54497111
Molybdenum, insulated wire, 50m, conductor diameter 0.125mm, insulation thickness 0.012mm, polyimide insulation, 99.95%
Mo
GF33272333
Molybdenum, mesh, 100x100mm, nominal aperture 0.44mm, wire diameter 0.07mm, 50 wires/inch, open area 67%, plain weave mesh
Mo
GF01979477
Molybdenum, mesh, 100x100mm, nominal aperture 0.8mm, wire diameter 0.18mm, 26 wires/inch, open area 67%, plain weave mesh
Mo