EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Synthesis and antitumor activity of new sulfonamide derivatives of thiadiazolo[3,2-a]pyrimidines.
PMID 21705114
Abstract
New series of sulfonamide derivatives of [1,3,4]thiadiazolo[3,2-a]pyrimidine were synthesized and investigated as antitumor agents. Some of the newly prepared compounds were tested for their inxa0vitro and inxa0vivo antitumor activities. Preliminary biological studies revealed that compounds 4c, 4f, and 4j exhibited the highest affinity to DNA, while compounds 4h,i, 6a-c, 8 and 12-14 exhibited moderate activity. Also, compounds 4j, 4f and 4c showed the highest percentage increase in lifespan of mice inoculated with Ehrlich ascites cells over 5-flurouracil (positive control). The detailed synthesis, spectroscopic and biological data are reported.