EMAIL THIS PAGE TO A FRIEND
Cancer biology & therapy
Heparan sulfate D-glucosaminyl 3-O-sulfotransferase-3B1, a novel epithelial-mesenchymal transition inducer in pancreatic cancer.
PMID 21709440
Abstract
Epithelial-mesenchymal transition (EMT) is a critical early event in tumorigenesis. The contribution of heparan sulfate (HS) to EMT has not been fully elucidated. HS D-glucosaminyl 3-O-sulfotransferase-3B1 (3-OST-3B1) participates in the final step of HS fine structure biosynthesis, whose involvement in cancer has yet to be determined. This study demonstrated that following treatment with trichostatin-A, a histone deacetylase inhibitor, 3-OST-3B1 gene expression was activated in the pancreatic cancer cell line, PANC-1. By chromatin immunoprecipitation analysis, permissive histone modifications including an increase in histone H3 lysine 9 monoactylation (H3 ac K9) but a decrease in methylated histone H3 (H3 me K9) were observed accompanying transcriptional activation of 3-OST-3B1. Functional, results revealed that increased 3-OST-3B1 levels were involved in the promotion of EMT processes. In vitro studies demonstrated that overexpression of 3-OST-3B1 in both pancreatic cancer cells and vascular endothelial cells could trigger an EMT-like phenotype as evidenced by the up-regulation of Snail at the mRNA and protein level, and its nuclear translocation. And 3-OST-3B1 appeared to be sufficient for the development of a more mesenchymal phenotype in vivo. Together, the results from this study unveiled a distinct function for 3-OST-3B1 as an EMT inducer in cancer and provided a link between histone modification and EMT modulation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CKOZFN10309 CompoZr® Knockout ZFN Kit, Human HS3ST3B1 (NM_006041)