EMAIL THIS PAGE TO A FRIEND
BMC cardiovascular disorders
High sensitive troponin T and heart fatty acid binding protein: novel biomarker in heart failure with normal ejection fraction? A cross-sectional study.
PMID 21729325
Abstract
High sensitive troponin T (hsTnT) and heart fatty acid binding protein (hFABP) are both markers of myocardial injury and predict adverse outcome in patients with systolic heart failure (SHF). We tested whether hsTnT and hFABP plasma levels are elevated in patients with heart failure with normal ejection fraction (HFnEF). We analyzed hsTnT, hFABP and N-terminal brain natriuretic peptide in 130 patients comprising 49 HFnEF patients, 51 patients with asymptomatic left ventricular diastolic dysfunction (LVDD), and 30 controls with normal diastolic function. Patients were classified to have HFnEF when the diagnostic criteria as recommended by the European Society of Cardiology were met. Levels of hs TnT and hFABP were significantly higher in patients with asymptomatic LVDD and HFnEF (both p < 0.001) compared to controls. The hsTnT levels were 5.6 [0.0-9.8] pg/ml in LVDD vs. 8.5 [3.9-17.5] pg/ml in HFnEF vs. <0.03 [< 0.03-6.4] pg/ml in controls; hFABP levels were 3029 [2533-3761] pg/ml in LVDD vs. 3669 [2918-4839] pg/ml in HFnEF vs. 2361 [1860-3081] pg/ml in controls. Furthermore, hsTnT and hFABP levels were higher in subjects with HFnEF compared to LVDD (p = 0.015 and p = 0.022). In HFnEF patients, hsTnT and hFABP are elevated independent of coronary artery disease, suggesting that ongoing myocardial damage plays a critical role in the pathophysiology. A combination of biomarkers and echocardiographic parameters might improve diagnostic accuracy and risk stratification of patients with HFnEF.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
RAB0657 Human FABP3 / Fatty Acid-binding Protein, Heart ELISA Kit