EMAIL THIS PAGE TO A FRIEND
Analytical chemistry
Direct detection of biologically significant thiols and disulfides with manganese(IV) chemiluminescence.
PMID 21732599
Abstract
The quantification of low-molecular mass thiols and disulfides involved in cellular redox processes is hindered by oxidation or degradation of analytes during conventional sample preparation steps (including deproteinization and derivatization). Researchers therefore seek techniques that minimize sample handling and permit direct detection of thiols and disulfides within a single chromatographic separation. We demonstrate a new HPLC procedure for these biologically important analytes that incorporates direct chemiluminescence detection with a manganese(IV) reagent. A mixture of seven thiols and disulfides (cysteine, N-acetylcysteine, homocysteine, glutathione (GSH), glutathione disulfide (GSSG), cystine, and homocystine) in their native forms were separated using a C18 column within 20 min. Detection limits for these analytes ranged from 5 × 10(-8) to 1 × 10(-7) M, and the precision for retention times and peak areas was excellent, with relative standard deviations of less than 0.3% and 2%, respectively. This approach was employed to determine two key biomarkers of oxidative stress, GSH and GSSG, in whole blood taken from 12 healthy volunteers. Samples were deproteinized, centrifuged, and diluted prior to analysis using a simple procedure that was shown to avoid significant artificial oxidation of GSH.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H6010
L-Homocystine, ≥98% (HPLC)
C8H16N2O4S2