EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Deoxyfluorination of phenols.
PMID 21736304
Abstract
An operationally simple ipso fluorination of phenols with a new deoxyfluorination reagent is presented.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
795291
Phenofluor solution, 0.1 M in toluene
C27H36F2N2