EMAIL THIS PAGE TO A FRIEND
Drug testing and analysis
The pharmacology and toxicology of the synthetic cathinone mephedrone (4-methylmethcathinone).
PMID 21755604
Abstract
Mephedrone (4-methylmethcathinone) is a synthetic cathinone that is used as a recreational drug. It has been available since 2007 but its availability and use increased significantly during 2009 and 2010. In this review article we will summarize the available literature on the sources, availability, and prevalence of the use of mephedrone. We will also discuss the pharmacology of mephedrone, the patterns of acute toxicity associated with its use, the reports of fatalities associated with its use, and the potential for mephedrone dependence.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
517925
4′-Methylpropiophenone, 90%, technical grade
C10H12O