EMAIL THIS PAGE TO A FRIEND
Analytical chemistry insights
The effect of varying short-chain alkyl substitution on the molar absorptivity and quantum yield of cyanine dyes.
PMID 21760707
Abstract
The effect of varying short-chain alkyl substitution of the indole nitrogens on the spectroscopic properties of cyanine dyes was examined. Molar absorptivities and fluorescence quantum yields were determined for a set of pentamethine dyes and a set of heptamethine dyes for which the substitution of the indole nitrogen was varied. For both sets of dyes, increasing alkyl chain length resulted in no significant change in quantum yield or molar absorptivity. These results may be useful in designing new cyanine dyes for analytical applications and predicting their spectroscopic properties.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
83701
Rhodamine 800, suitable for fluorescence
C26H26ClN3O5