EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
The discovery and development of inhibitors of fatty acid amide hydrolase (FAAH).
PMID 21764305
Abstract
A summary of the discovery and advancement of inhibitors of fatty acid amide hydrolase (FAAH) is presented.