EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Synthesis of oligoribonucleotides with phosphonate-modified linkages.
PMID 21769370
Abstract
Solid phase synthesis of phosphonate-modified oligoribonucleotides using 2'-O-benzoyloxymethoxymethyl protected monomers is presented in both 3'→5' and 5'→3' directions. Hybridisation properties and enzymatic stability of oligoribonucleotides modified by regioisomeric 3'- and 5'-phosphonate linkages are evaluated. The introduction of the 5'-phosphonate units resulted in moderate destabilisation of the RNA/RNA duplexes (ΔT(m)-1.8 °C/mod.), whereas the introduction of the 3'-phosphonate units resulted in considerable destabilisation of the duplexes (ΔT(m)-5.7 °C/mod.). Molecular dynamics simulations have been used to explain this behaviour. Both types of phosphonate linkages exhibited remarkable resistance in the presence of ribonuclease A, phosphodiesterase I and phosphodiesterase II.