EMAIL THIS PAGE TO A FRIEND
Acta neuropathologica
Enhancement of endogenous neurogenesis in ephrin-B3 deficient mice after transient focal cerebral ischemia.
PMID 21779764
Abstract
Cerebral ischemia stimulates endogenous neurogenesis. However, the functional relevance of this phenomenon remains unclear because of poor survival and low neuronal differentiation rates of newborn cells. Therefore, further studies on mechanisms regulating neurogenesis under ischemic conditions are required, among which ephrin-ligands and ephrin-receptors (Eph) are an interesting target. Although Eph/ephrin proteins like ephrin-B3 are known to negatively regulate neurogenesis under physiological conditions, their role in cerebral ischemia is largely unknown. We therefore studied neurogenesis, brain injury and functional outcome in ephrin-B3(-/-) (knockout) and ephrin-B3(+/+) (wild-type) mice submitted to cerebral ischemia. Induction of stroke resulted in enhanced cell proliferation and neuronal differentiation around the lesion site of ephrin-B3(-/-) compared to ephrin-B3(+/+) mice. However, prominent post-ischemic neurogenesis in ephrin-B3(-/-) mice was accompanied by significantly increased ischemic injury and motor coordination deficits that persisted up to 4xa0weeks. Ischemic injury in ephrin-B3(-/-) mice was associated with a caspase-3-dependent activation of the signal transducer and activator of transcription 1 (STAT1). Whereas inhibition of caspase-3 had no effect on brain injury in ephrin-B3(+/+) animals, infarct size in ephrin-B3(-/-) mice was strongly reduced, suggesting that aggravated brain injury in these animals might involve a caspase-3-dependent activation of STAT1. In conclusion, post-ischemic neurogenesis in ephrin-B3(-/-) mice is strongly enhanced, but fails to contribute to functional recovery because of caspase-3-mediated aggravation of ischemic injury in these animals. Our results suggest that ephrin-B3 might be an interesting target for overcoming some of the limitations of further cell-based therapies in stroke.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
19899
CHAPS 100 mM solution
C32H58N2O7S