EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Metastable linkage isomerism in [Ni(Et4dien)(NO2)2]: a combined thermal and photocrystallographic structural investigation of a nitro/nitrito interconversion.
PMID 21780258
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
424072
N,N,N′,N′-Tetraethyldiethylenetriamine, technical grade, 90%
C12H29N3