EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Synthesis and topoisomerase I inhibitory activity of a novel diazaindeno[2,1-b]phenanthrene analogue of Lamellarin D.
PMID 21783369
Abstract
A novel 5-oxa-6a,8-diazaindeno[2,1-b]phenanthren-7-one scaffold was designed and synthesized as an active analogue of the cytotoxic marine alkaloid Lamellarin D. The design was based on molecular modeling of the site of interaction of Lamellarin D with DNA-topoisomerase I cleavable complex, whereas the synthesis capitalized on a simple Friedel-Crafts cyclization of indole to a β-carbolinone nucleus. The product exhibited topoisomerase I poisoning activity and submicromolar cytotoxicity on human non-small cell lung cancer H460 cell line.
Related Materials