EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Synthesis, gp120 binding and anti-HIV activity of fatty acid esters of 1,1-linked disaccharides.
PMID 21783371
Abstract
Inspired by the anti-human immunodeficiency virus (HIV) activity of analogues of β-galactosylceramide (GalCer), a set of mono- and di-saccharide fatty acid esters were designed as GalCer mimetics and their binding to the V3 loop peptide of HIV-1 and anti-HIV activity evaluated. 1,1-linked Gal-Man and Glu-Man disaccharides with an ester on the Man subunit bound the V3 loop peptide and inhibited HIV infectivity in single round infection assays with the TZM-bl cell line. IC(50)'s were in the 50 μM range with no toxicity to the cells at concentrations up to 200 μM. These compounds appear to inhibit virus entry at early steps in viral infection since they were inactive if added post viral entry. Although these compounds were found to bind to the V3 loop peptide of gp120, it is not clear that this interaction is responsible for their anti-HIV activity because the relative binding affinity of closely related analogues did not correlate with their antiviral behavior. The low cytotoxicity of these 1,1-linked disaccharide fatty acid esters, combined with the easy accessibility to structurally diverse analogues make these molecules attractive leads for new topical anti-viral agents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M1765
1-Lauroyl-rac-glycerol, ≥99%
C15H30O4