EMAIL THIS PAGE TO A FRIEND
Veterinary parasitology
Antioxidant enzymes in erythrocytes from goats seropositive to the sheep nose bot fly (Oestrus ovis L., Diptera: Oestridae) infection.
PMID 21802210
Abstract
Oestrus ovis (Diptera: Oestridae) causes an important cosmopolitan parasitosis of the nasal and sinusal cavities of sheep and goats called oestrosis. Our objective was to analyze the participation of erythrocytes in the antioxidant system in goats seropositive to O. ovis infection under field conditions. Fifty female goats naturally exposed to O. ovis infection from Baja California Sur, México, were blood-sampled. Erythrocytic intracellular content was obtained from blood plasma. Oestrosis serodiagnosis was determined by ELISA. Protein, hemoglobin (Hb), superoxide dismutase (SOD), mieloperoxidase (MPO), catalase (CAT), glutathione-S-transferase (GST), and lipid peroxidation in erythrocytes were determined in both seropositive and seronegative goats. Overall seroprevalence of O. ovis infection in goats was 56%. Positive significant (P<0.05) associations were observed among systemic IgG level and protein (0.34), hemoglobin (0.43), SOD (0.32), and MPO (0.41) in erythrocytes. Protein and hemoglobin concentrations, as well as SOD and MPO activities in erythrocytes were found significantly higher (P<0.05) in seropositive than in seronegative goats. By contrast, enzymatic activities of CAT and GST and lipid peroxidation values were similar in seropositive and seronegative groups. In conclusion, there was a systemic stimulation of Reactive Oxygen Species which was efficiently scavenged by erythrocytic antioxidant enzymes in goats seropositive to O. ovis infection.