EMAIL THIS PAGE TO A FRIEND
Chembiochem : a European journal of chemical biology
Intracellular detection of cytosine incorporation in genomic DNA by using 5-ethynyl-2'-deoxycytidine.
PMID 21805552
Abstract
5-Ethynyl-2'-deoxycytidine triphosphate (EdCTP) was synthesized as a probe to be used in conjunction with fluorescent labeling to facilitate the analysis of the in vivo dynamics of DNA-centered processes (DNA replication, repair and cytosine demethylation). Kinetic analysis showed that EdCTP is accepted as a substrate by Klenow exo(-) and DNA polymerase β. Incorporation of 5-ethynyl-2'-deoxycytidine (EdC) into DNA by these enzymes is, at most, modestly less efficient than native dC. EdC-containing DNA was visualized by using a click reaction with a fluorescent azide, following polymerase incorporation and T4 DNA ligase mediated ligation. Subsequent experiments in mouse male germ cells and zygotes demonstrated that EdC is a specific and reliable reporter of DNA replication, in vivo.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T511307
5-Ethynyl-2′-deoxycytidine, (EdC), AldrichCPR
C11H13N3O4