EMAIL THIS PAGE TO A FRIEND
Organic letters
Stereoselective synthesis of substituted 1,3-oxazolidines via Pd-catalyzed carboamination reactions of O-vinyl-1,2-amino alcohols.
PMID 21812450
Abstract
The stereoselective synthesis of 2,4- and 2,5-disubstituted 1,3-oxazolidines is accomplished via Pd-catalyzed carboamination of O-vinyl-1,2-amino alcohol derivatives. The transformations generate cis-disubstituted products with good to excellent diastereoselectivity, and enantiomerically enriched substrates are converted without loss of optical purity. In addition to yielding synthetically useful products that are difficult to generate with existing methods, these transformations illustrate that electron-rich enol ethers are viable substrates for alkene carboamination processes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
637998
Vinylboronic anhydride pyridine complex, 95%
C6H9B3O3 · C5H5N