EMAIL THIS PAGE TO A FRIEND
Parasitology
Leishmania donovani: proteasome-mediated down-regulation of methionine adenosyltransferase.
PMID 21813028
Abstract
Methionine adenosyltransferase (MAT) is an important enzyme for metabolic processes, to the extent that its product, S-adenosylmethionine (AdoMet), plays a key role in trans-methylation, trans-sulphuration and polyamine synthesis. Previous studies have shown that a MAT-overexpressing strain of Leishmania donovani controls AdoMet production, keeping the intracellular AdoMet concentration at levels that are compatible with cell survival. This unexpected result, together with the fact that MAT activity and abundance changed with time in culture, suggests that different regulatory mechanisms acting beyond the post-transcriptional level are controlling this protein. In order to gain an insight into these mechanisms, several experiments were carried out to explain the MAT abundance during promastigote cell growth. Determination of MAT turnover in cycloheximide (CHX)-treated cultures resulted in a surprising 5-fold increase in MAT turnover compared to CHX-untreated cultures. This increase agrees with a stabilization of the MAT protein, whose integrity was maintained during culture. The presence of proteasome inhibitors, namely MG-132, MG-115, epoxomycin and lactacystin in the culture medium prevented MAT degradation in both MAT-overexpressing and 'mock-transfected' leishmanial strains. The role of the ubiquitin (Ub) pathway in MAT down-regulation was supported using immunoprecipitation experiments. Immunoprecipitated MAT cross-reacted with anti-Ub antibodies, which provides evidence of a proteasome-mediated down-regulation of the leishmanial MAT abundance.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SCP0005 MG-115 / ASD 4955, ≥97% (HPLC)
C25H39N3O5