EMAIL THIS PAGE TO A FRIEND
Journal of physics. Condensed matter : an Institute of Physics journal
Study of the electronic structure at the interface between fluorene-1-carboxylic acid molecules and Cu(110).
PMID 21828626
Abstract
The interface electronic properties of fluorene-1-carboxylic acid (FC-1) adsorbed on Cu(110) have been studied by ultraviolet photoemission spectroscopy (UPS) and first-principles calculations. Both the molecular orbitals and the Cu valence band are significantly modified upon adsorption. FC-1 is chemically bonded to Cu(110) through charge donation and back donation involving the lowest unoccupied molecular orbital (LUMO) and the highest occupied molecular orbital (HOMO) of the molecule. An observed reduction of the work function can be attributed to the adsorption induced charge redistribution, and the positive interface dipole.