EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Structure-based design of new DHFR-based antibacterial agents: 7-aryl-2,4-diaminoquinazolines.
PMID 21831637
Abstract
Dihydrofolate reductase (DHFR) inhibitors such as trimethoprim (TMP) have long played a significant role in the treatment of bacterial infections. Not surprisingly, after decades of use there is now bacterial resistance to TMP and therefore a need to develop novel antibacterial agents with expanded spectrum including these resistant strains. In this study, we investigated the optimization of 2,4-diamnoquinazolines for antibacterial potency and selectivity. Using structure-based drug design, several 7-aryl-2,4-diaminoquinazolines were discovered that have excellent sub-100 picomolar potency against bacterial DHFR. These compounds have good antibacterial activity especially on gram-positive pathogens including TMP-resistant strains.