EMAIL THIS PAGE TO A FRIEND
Development (Cambridge, England)
Cell cycle arrest in node cells governs ciliogenesis at the node to break left-right symmetry.
PMID 21831921
Abstract
Cilia at the node generate a leftward fluid flow that breaks left-right symmetry. However, the molecular mechanisms that regulate ciliogenesis at the node are largely unknown. Here, we show that the epiblast-specific deletion of the gene encoding the BMP type 1 receptor (Acvr1) compromised development of nodal cilia, which results in defects in leftward fluid flow and, thus, abnormalities in left-right patterning. Acvr1 deficiency in mouse embryonic fibroblasts (MEFs) resulted in severe defects in their quiescence-induced primary cilia. Although the induction of quiescence in wild-type MEFs leads to an increase in the level of the cyclin-dependent kinase inhibitor p27(Kip1) and to rapid p27(Kip1) phosphorylation on Ser(10), MEFs deficient in Acvr1 show a reduction in both p27(Kip1) protein levels and in p27(Kip1) Ser(10) phosphorylation. The observed defects in cilium development were rescued by the introduction of p27(Kip1) into Acvr1-deficient MEFs, implying that BMP signaling positively controls p27(Kip1) stability in the G0 phase via p27(Kip1) Ser(10) phosphorylation, which is a prerequisite for induction of primary cilia. Importantly, in control embryos, p27(Kip1) protein is clearly present and strongly phosphorylated on Ser(10) in cells on the quiescent ventral surface of the node. By contrast, the corresponding cells in the node of Acvr1 mutant embryos were proliferative and showed a dramatic attenuation in both p27(Kip1) protein levels and phosphorylation on Ser(10). Our data suggest that cell quiescence controlled by BMP signaling via ACVR1 is required for transient formation of nodal cilia, and provide insight into the fundamental question of how the node represents the mechanistic `node' that regulates the development of left-right symmetry in vertebrates.
Related Materials