EMAIL THIS PAGE TO A FRIEND
The Journal of investigative dermatology
Clinical, molecular, and cellular features of non-Puerto Rican Hermansky-Pudlak syndrome patients of Hispanic descent.
PMID 21833017
Abstract
Hermansky-Pudlak syndrome (HPS) is an autosomal recessive condition characterized by a bleeding diathesis and hypopigmentation of the skin, hair, and eyes. Some HPS patients develop other complications such as granulomatous colitis and/or fatal pulmonary fibrosis. Eight genes have been associated with this condition, resulting in subtypes HPS-1 through HPS-8. The HPS gene products are involved in the biogenesis of specialized lysosome-related organelles such as melanosomes and platelet delta granules. HPS1 and HPS4 form a stable complex named biogenesis of lysosome-related organelles complex (BLOC)-3, and patients with BLOC-3 or AP-3 deficiency develop pulmonary fibrosis. Therefore, it is important to subtype each HPS patient. HPS type 1 (HPS-1) occurs frequently on the island of Puerto Rico because of a founder mutation. Here, we describe seven mutations, six of which, to our knowledge, are previously unreported in the HPS1, HPS4, and HPS5 genes among patients of Mexican, Uruguayan, Honduran, Cuban, Venezuelan, and Salvadoran ancestries. Our findings demonstrate that the diagnosis of HPS should be considered in Hispanic patients with oculocutaneous albinism and bleeding symptoms. Moreover, such patients should not be assumed to have the HPS-1 subtype typical of northwest Puerto Rican patients. We recommend molecular HPS subtyping in such cases, as it may have significant implications for prognosis and intervention.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CKOZFN10279 CompoZr® Knockout ZFN Kit, Human HPS1 (NM_000195)