EMAIL THIS PAGE TO A FRIEND
International journal of molecular medicine
Inhibitory effects of vitamin K₃ derivatives on DNA polymerase and inflammatory activity.
PMID 21837358
Abstract
Previously, we reported that vitamin K₃ (menadione, 2-methyl-1,4-naphthoquinone) (compound 2) inhibits the activity of human mitochondrial DNA polymerase γ (pol γ). In this study, we investigated the inhibitory effects (IEs) of vitamin K3 and its derivatives, such as 1,4-naphthoquinone (compound 1) and 1,2-dimethyl-1,4-naphthoquinone (compound 3), on the activity of mammalian pols. Among compounds 1-3 (10 µM for each), compound 1 was the strongest inhibitor of mammalian pols α and λ, which belong to the B and X pol families, respectively, whereas compound 2 was the strongest inhibitor of human pol γ, a family A pol. However, these compounds did not affect the activity of human pol κ, a family Y pol. As we previously found a positive relationship between pol λ inhibition and anti-inflammatory action, we examined whether these vitamin K₃ derivatives are able to inhibit inflammatory responses. Among the three compounds tested, compound 1 caused the greatest reduction in 12-O-tetradecanoylphorbol-13-acetate (TPA)-induced acute inflammation in mouse ears. In addition, in a cell culture system using RAW264.7 mouse macrophages, compound 1 displayed the strongest suppression of tumor necrosis factor (TNF)-α production induced by lipopolysaccharide (LPS). In an in vivo mouse model of LPS-evoked acute inflammation, the intraperitoneal injection of compound 1 into mice suppressed TNF-α production in their peritoneal macrophages and serum. In an in vivo colitis mouse model induced using dextran sulfate sodium (DSS), the vitamin K₃ derivatives markedly suppressed DSS-evoked colitis. In conclusion, this study has identified several vitamin K₃ derivatives, such as compound 1, that are promising anti-inflammatory candidates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M5625
Menadione, crystalline
C11H8O2