EMAIL THIS PAGE TO A FRIEND
Evidence-based complementary and alternative medicine : eCAM
Novel lipolytic enzymes identified from metagenomic library of deep-sea sediment.
PMID 21845199
Abstract
Metagenomic library was constructed from a deep-sea sediment sample and screened for lipolytic activity. Open-reading frames of six positive clones showed only 33-58% amino acid identities to the known proteins. One of them was assigned to a new group while others were grouped into Families I and V or EstD Family. By employing a combination of approaches such as removing the signal sequence, coexpression of chaperone genes, and low temperature induction, we obtained five soluble recombinant proteins in Escherichia coli. The purified enzymes had optimum temperatures of 30-35°C and the cold-activity property. Among them, one enzyme showed lipase activity by preferentially hydrolyzing p-nitrophenyl palmitate and p-nitrophenyl stearate and high salt resistance with up to 4 M NaCl. Our research demonstrates the feasibility of developing novel lipolytic enzymes from marine environments by the combination of functional metagenomic approach and protein expression technology.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N9876
4-Nitrophenyl butyrate, ≥98%
C10H11NO4