EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
An easily accessible isoindigo-based polymer for high-performance polymer solar cells.
PMID 21848297
Abstract
A new, low-band-gap alternating copolymer consisting of terthiophene and isoindigo has been designed and synthesized. Solar cells based on this polymer and PC(71)BM show a power conversion efficiency of 6.3%, which is a record for polymer solar cells based on a polymer with an optical band gap below 1.5 eV. This work demonstrates the great potential of isoindigo moieties as electron-deficient units for building donor-acceptor-type polymers for high-performance polymer solar cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
802123
6,6′-Dibromoisoindigo
C16H8Br2N2O2