EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Synthesis and glycosidase inhibitory profiles of functionalised morpholines and oxazepanes.
PMID 21862336
Abstract
In this work libraries of morpholines and oxazepanes have been prepared via the reductive amination reaction between dialdehydes, derived from carbohydrates, and a range of amines. In this way, functionalised morpholines and oxazepanes have been prepared that include N-alkylated derivatives, disaccharide analogues, and ester containing derivatives. The abilities of these functionalised morpholines and oxazepanes to inhibit a broad panel of glycosidase enzymes, that are associated with a range of diseases, have been probed and in this way new inhibitors of a range of glycosidases, but particularly β-d-galactosidase derived from Bovine kidney, have been discovered. N-Alkyl morpholines demonstrated the best inhibition profiles for this enzyme and derivatives (15a)-(15d) acted as non-competitive inhibitors with IC(50) values of 55.1-88.6 μM. Within this study, some preliminary structure-activity relationships are proposed, and it is demonstrated that N-substituted morpholines display better inhibitory profiles for the enzymes analysed than any of the N-substituted oxazepanes.