EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Synthesis and antioxidant activity of oxazolyl/thiazolylsulfonylmethyl pyrazoles and isoxazoles.
PMID 21864949
Abstract
A new class of oxazolyl/thiazolylsulfonylmethyl pyrazoles (10-13) and isoxazoles (14, 15) were prepared from the synthetically vulnerable intermediate E-styrylsulfonylacetic acid methyl ester (1) and studied their antioxidant activity.