EMAIL THIS PAGE TO A FRIEND
Nature
Forum. Chemical engineering: Fuel for debate.
PMID 21866147
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6288
Cellulose, fibers, (medium)