EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis and in vitro antiprotozoal activities of 5-phenyliminobenzo[a]phenoxazine derivatives.
PMID 21868222
Abstract
A series of 5-phenyliminobenzo[a]phenoxazine derivatives were synthesized. The in vitro antiprotozoal activities were evaluated against Plasmodium falciparum K1, Trypanosoma cruzi, Leishmania donovani and Trypanosoma brucei rhodesiense. N,N-Diethyl-5-((4-methoxyphenyl)imino)-5H-benzo[a]phenoxazin-9-amine shows IC(50)=0.040 μmol L(-1) with a selective index of 1425 against Plasmodium falciparum K1.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
263893
Phenoxazine, ≥99%, purified by sublimation
C12H9NO
P14858
Phenoxazine, 97%
C12H9NO