EMAIL THIS PAGE TO A FRIEND
BMC genomics
A eukaryotic-type signalling system of Pseudomonas aeruginosa contributes to oxidative stress resistance, intracellular survival and virulence.
PMID 21880152
Abstract
The genome of Pseudomonas aeruginosa contains at least three genes encoding eukaryotic-type Ser/Thr protein kinases, one of which, ppkA, has been implicated in P. aeruginosa virulence. Together with the adjacent pppA phosphatase gene, they belong to the type VI secretion system (H1-T6SS) locus, which is important for bacterial pathogenesis. To determine the biological function of this protein pair, we prepared a pppA-ppkA double mutant and characterised its phenotype and transcriptomic profiles. Phenotypic studies revealed that the mutant grew slower than the wild-type strain in minimal media and exhibited reduced secretion of pyoverdine. In addition, the mutant had altered sensitivity to oxidative and hyperosmotic stress conditions. Consequently, mutant cells had an impaired ability to survive in murine macrophages and an attenuated virulence in the plant model of infection. Whole-genome transcriptome analysis revealed that pppA-ppkA deletion affects the expression of oxidative stress-responsive genes, stationary phase σ-factor RpoS-regulated genes, and quorum-sensing regulons. The transcriptome of the pppA-ppkA mutant was also analysed under conditions of oxidative stress and showed an impaired response to the stress, manifested by a weaker induction of stress adaptation genes as well as the genes of the SOS regulon. In addition, expression of either RpoS-regulated genes or quorum-sensing-dependent genes was also affected. Complementation analysis confirmed that the transcription levels of the differentially expressed genes were specifically restored when the pppA and ppkA genes were expressed ectopically. Our results suggest that in addition to its crucial role in controlling the activity of P. aeruginosa H1-T6SS at the post-translational level, the PppA-PpkA pair also affects the transcription of stress-responsive genes. Based on these data, it is likely that the reduced virulence of the mutant strain results from an impaired ability to survive in the host due to the limited response to stress conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E0502 Elastin–Congo red, elastase substrate
12107C
Fetal Bovine Serum, USA origin, γ-irradiated by SER-TAIN® process, sterile-filtered, suitable for cell culture
12303C
Fetal Bovine Serum, USDA approved countries, sterile-filtered, cell culture tested
12003C
Fetal Bovine Serum, Australia origin, USDA approved, sterile-filtered, suitable for cell culture
12007C
Fetal Bovine Serum, Australia origin, γ-irradiated, sterile-filtered, suitable for cell culture
12203C
Fetal Bovine Serum, New Zealand origin, sterile-filtered, suitable for cell culture
F0926
Fetal Bovine Serum, USDA approved, sterile-filtered, suitable for cell culture
12306C
Fetal Bovine Serum, non-USA origin, from USDA approved countries, Heat Inactivated, sterile-filtered, suitable for cell culture
12006C
Fetal Bovine Serum, Australia origin, USDA approved, Heat Inactivated, suitable for cell culture
12207C
Fetal Bovine Serum, New Zealand origin, cell culture tested
F9423
Fetal Bovine Serum, Australia origin, suitable for, USDA approved, sterile-filtered, suitable for cell culture, suitable for hybridoma
F3297
Fetal Bovine Serum, Australia origin, γ-irradiated, cell culture tested
12105C
Fetal Bovine Serum, Dialyzed by ultrafiltration against 0.15M NaCl, USA origin, sterile-filtered, cell culture tested
12103C
Fetal Bovine Serum, USA origin, cell culture tested
12106C
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture
F6765
Fetal Bovine Serum, USA origin, Charcoal Stripped, sterile-filtered, suitable for cell culture
F7305
Fetal Bovine Serum, Mouse Embryonic Stem Cell Screened, suitable for cell culture, sterile-filtered, USDA Approved
P2893
JumpStart Taq ReadyMix, Complete optimized reagent for hot-start PCR at 2X concentration