EMAIL THIS PAGE TO A FRIEND
Investigational new drugs
Phase I and pharmacokinetic study of dasatinib and cetuximab in patients with advanced solid malignancies.
PMID 21881918
Abstract
Combined inhibition of epidermal growth factor receptor (EGFR) and Src family kinases (SFK) may lead to improved therapeutic effects. We evaluated the combination of dasatinib, an inhibitor of SFK and other kinases, and cetuximab, an anti-EGFR monoclonal antibody. Patients with advanced solid malignancies received cetuximab intravenously on a standard weekly schedule and dasatinib orally, once daily at 3 dose levels: (1) 100 mg, (2) 150 mg, (3) 200 mg. Pharmacokinetic and pharmacodynamic studies of dasatinib were performed prior to starting cetuximab and following 14 days of treatment. Twenty-five patients (3 dose level 1; 19 dose level 2; 3 dose level 3) were initially treated. Three patients developed dose-limiting toxicities: 1 at dose level 2 (headache) and 2 at dose level 3 (headache, nausea). Grade 3-4 toxicities in more than 2 patients included: dyspnea (4), vomiting (4), nausea (3), hypersensitivity reactions (3), headache (3) and anemia (3). Twenty-one patients developed headache (8 grade 1; 10 grade 2), which occurred after the loading of cetuximab and lasted 1-3 days. Six additional patients were treated with dasatinib starting 3 days after the loading dose of cetuximab; none developed headache after dasatinib. Dasatinib pharmacokinetics and a transient decrease in SFK PY416 levels in peripheral blood mononuclear cells were not altered by cetuximab. Patients with higher plasma TGF-alpha levels had worse progression-free survival. Dasatinib 150 mg once daily plus weekly cetuximab is recommended for phase II studies. Early-onset headache was ameliorated by starting dasatinib after cetuximab.