EMAIL THIS PAGE TO A FRIEND
Physical chemistry chemical physics : PCCP
J-coupling constants for a trialanine peptide as a function of dihedral angles calculated by density functional theory over the full Ramachandran space.
PMID 21897927
Abstract
We present 13 (3)J, seven (2)J and four (1)J coupling constants (24 in all) calculated using B3LYP/D95** as a function of the φ and ψ Ramachandran dihedral angles of the acetyl(Ala)(3)NH(2) capped trialanine peptide over the entire Ramachandran space. With the exception of three of these J couplings, all show significant dependence upon both dihedral angles. For each J coupling considered, a two dimensional grid with respect to φ and ψ angles can be used to interpolate the values for any pair of φ and ψ values. Such simple interpolation is shown to be very accurate. Most of these calculated J couplings should prove useful for improving the accuracy of the determination of peptide and protein structures from NMR measurements in solution over that provided by the common procedure of treating the J couplings as functions of a single dihedral angle by means of Karplus-type fittings.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9627
Ala-Ala-Ala
C9H17N3O4